ContactUsLinkComponent
Kit exhaust cabinet hardware exhaust purge connection for the sps 3 dust cover. 5 mm (2 in.) diameter outlet. extracts corrosive or solvent fumes when connected to an exhaust system. 1 * 1 items
Catalog # VARI9910130600
Supplier:  VARIAN
Kit exhaust cabinet hardware exhaust purge connection for the sps 3 dust cover. 5 mm (2 in.) diameter outlet. extracts corrosive or solvent fumes when connected to an exhaust system. 1 * 1 items
Catalog # VARI9910130600
Supplier:  VARIAN
Frequently Bought Together