Orders LinkContactUsLinkComponent
Vials
Vials
Vials, with write on label, 2,00 ml, Amber, No
Catalog # HEWL5190-6120
Supplier:   Agilent
Vials
Catalog # HEWL5190-6120
Supplier:  Agilent
Vials
Vials
Vials, with write on label, 2,00 ml, Amber, No
Price
11 560,00 €
Log in for availability
1 * 50.000 ST
Product Details & Documents
Specifications
  • Cap included:
    No
  • Capacity:
    2,00 ml
  • Colour:
    Amber
  • Description:
    Vials, with write on label
  • Seq:
    22