Order Entry
Austria
Orders LinkContactUsLinkComponent
Detectable plaster rolls

Detectable plaster rolls

Detectable plaster roll, 5 m, 5000×60 mm
Catalog # SOEH1009998
Supplier:   SOEHNGEN
Detectable plaster rolls
Catalog # SOEH1009998
Supplier:  SOEHNGEN
Detectable plaster rolls

Detectable plaster roll, 5 m, 5000×60 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

These detectable plasters are used to cover small skin lesions. The aluminium strip under the wound pad make the plasters detectable by all conventional metal detectors. Ideal for certain areas of production, in particular the food industry.

  • Skin-friendly and reliable acrylic adhesive
  • Water resistant, hypoallergenic and breathable
  • Blue colour ensures easy visual recognition
Specifications
  • Description:
    Detectable plaster roll, 5 m
  • L×W:
    5000×60 mm