ContactUsLinkComponent
Spectromelt® C 15 MP Micropowder grinding and tabletting aid for X-ray fluorescence spectroscopy 1 * 1 kg
Catalog # 1.17074.1000
Supplier:  Merck
Spectromelt® C 15 MP Micropowder grinding and tabletting aid for X-ray fluorescence spectroscopy 1 * 1 kg
Catalog # 1.17074.1000
Supplier:  Merck
Frequently Bought Together