ContactUsLinkComponent
Ultra Polar C19 x 1.8µm particle size x 120Å pore size, 100mm length x 2.1mm ID, Ultrahigh pressure hardware ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2UPOL-18M02U
Ultra Polar C19 x 1.8µm particle size x 120Å pore size, 100mm length x 2.1mm ID, Ultrahigh pressure hardware ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2UPOL-18M02U
Frequently Bought Together