ContactUsLinkComponent
Ultra HILIC FL x 1.8µm particle size x 120Å pore size, 150mm length x 2.1mm ID, Ultrahigh pressure hardware ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2UHL8-18N02U
undefined
Ultra HILIC FL x 1.8µm particle size x 120Å pore size, 150mm length x 2.1mm ID, Ultrahigh pressure hardware ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2UHL8-18N02U
Frequently Bought Together