ContactUsLinkComponent
C18G x 5µm particle size x 300Å pore size, 150mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-05N02S
C18G x 5µm particle size x 300Å pore size, 150mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-05N02S
Frequently Bought Together