ContactUsLinkComponent
C18G x 5µm particle size x 300Å pore size, 100mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-05M46S
C18G x 5µm particle size x 300Å pore size, 100mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C184-05M46S
Frequently Bought Together