ContactUsLinkComponent
Silica x 3µm particle size x 120Å pore size, 100mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2SIL-03M02S
Silica x 3µm particle size x 120Å pore size, 100mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2SIL-03M02S
Frequently Bought Together