ContactUsLinkComponent
C18Q x 5µm particle size x 100Å pore size, 250mm length x 1.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-05R01S
C18Q x 5µm particle size x 100Å pore size, 250mm length x 1.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C183-05R01S
Frequently Bought Together