ContactUsLinkComponent
Phenyl Hexylx 3µm particle size x 120Å pore size, 150mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2NH4-03N46P
Phenyl Hexylx 3µm particle size x 120Å pore size, 150mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2NH4-03N46P
Frequently Bought Together