ContactUsLinkComponent
HILIC RP x 5µm particle size x 120Å pore size, 250mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL6-05R02S
undefined
HILIC RP x 5µm particle size x 120Å pore size, 250mm length x 2.1mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2HL6-05R02S
Frequently Bought Together