ContactUsLinkComponent
Chiral CHA x 10µm particle size x 1000Å pore size, 250mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CHA-10R46S
undefined
Chiral CHA x 10µm particle size x 1000Å pore size, 250mm length x 4.6mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2CHA-10R46S
Frequently Bought Together