ContactUsLinkComponent
C18G x 3µm particle size x 120Å pore size, 150mm length x 1.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
  SGEA2C185-03N01S
C18G x 3µm particle size x 120Å pore size, 150mm length x 1.0mm ID, Stainless steel ProteCol HPLC Column 1 * 1 items
  SGEA2C185-03N01S
Frequently Bought Together