Orders LinkContactUsLinkComponent
Stopcocks, Galtek®
Stopcocks, Galtek®
Inlet 1/4" Flaretek® flared tube, outlet 1/4" Flaretek® flared tube, length 85,9 mm
Catalog # ENTEPVA4-T4-2FN-1
Supplier:   ENTEGRIS
Stopcocks, Galtek®
Catalog # ENTEPVA4-T4-2FN-1
Supplier:  ENTEGRIS
Stopcocks, Galtek®
Inlet 1/4" Flaretek® flared tube, outlet 1/4" Flaretek® flared tube, length 85,9 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Two- or three way, ¹/₈" orifice.
  • All wetted parts are PFA and PTFE.
  • Small compact design provides minimal internal volume.
  • Several connection sizes available to choose from
Specifications
  • Description:
    Inlet 1/4" Flaretek® flared tube, outlet 1/4" Flaretek® flared tube, length 85,9 mm