ContactUsLinkComponent
BOUFFANT CAP COLOR: BLUE SIZE: 58CM DESCRIPTION: CLEANED, LATEXFREE ELASTIC MATERIAL: POLYPROPYLENE THICKNESS: 10-12 G/M PACKAGING: 100 PCS/DOUBLE POLYBAG, 1000 PCS/CARTON 1 * 1.000 items
Catalog # GUNA130923
Supplier:  VWR Collection
BOUFFANT CAP COLOR: BLUE SIZE: 58CM DESCRIPTION: CLEANED, LATEXFREE ELASTIC MATERIAL: POLYPROPYLENE THICKNESS: 10-12 G/M PACKAGING: 100 PCS/DOUBLE POLYBAG, 1000 PCS/CARTON 1 * 1.000 items
Catalog # GUNA130923
Supplier:  VWR Collection
Frequently Bought Together