ContactUsLinkComponent
Überziehstiefel BDBO 18 (455mm) High Durability Cleanroom Overbshoes 102gsm Non-woven, spun bonded Polypropylene/Cast Polyethylene laminate STORAGE INSTRUCTIONS: Store in a dry, cool place (40C) away from direct sunlight and fluorescent li
Catalog # NITRBDBO18455MM
Supplier:  Nitritex
Überziehstiefel BDBO 18 (455mm) High Durability Cleanroom Overbshoes 102gsm Non-woven, spun bonded Polypropylene/Cast Polyethylene laminate STORAGE INSTRUCTIONS: Store in a dry, cool place (40C) away from direct sunlight and fluorescent li
Catalog # NITRBDBO18455MM
Supplier:  Nitritex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.