Nitril HS AMBI Indigo BNPLS Colour: Indigo Length: 300mm (12) Material: Nitrile Surface: Textured Sterile: Sterile Other Features: Powder-free, beaded cuff. Protein Level: Nitrile contains no natural latex proteins. Sterilisation Method: Gamma irr
Catalog # NITRBNPLS
Supplier: Nitritex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.