Stopcocks, Galtek®
All three ports 1/4" integral ferrule, T-bore
Catalog # ENTEPVT4-T4-T4-2
Supplier: ENTEGRIS
Jump to:
Product Details & Documents
Two- or three way, ¹/₈" orifice.
- All wetted parts are PFA and PTFE.
- Small compact design provides minimal internal volume.
- Several connection sizes available to choose from
Specifications
- Bezeichnung:All three ports 1/4" integral ferrule, T-bore