Orders LinkContactUsLinkComponent
Forceps
Forceps
Curved, tapered thin tip, 127 mm
Catalog # ENTEC14-0315
Supplier:   ENTEGRIS
Forceps
Catalog # ENTEC14-0315
Supplier:  ENTEGRIS
Forceps
Curved, tapered thin tip, 127 mm
Price
Inquire For Price
Log in for availability
1 * 4 ST
Product Details & Documents
ETFE, white. Durable forceps for substrate handling.
  • Different tip styles available
  • Packaged in protective cases
Specifications
  • Type:
    Curved, tapered thin tip
  • Length:
    127 mm
  • Seq:
    04