Order Entry
Austria
Orders LinkContactUsLinkComponent
Forceps

Forceps

Straight, 0,3 mm thick tip, 124 mm
Catalog # ENTEC10-0315
Supplier:   ENTEGRIS
Forceps
Catalog # ENTEC10-0315
Supplier:  ENTEGRIS
Forceps

Straight, 0,3 mm thick tip, 124 mm

Price
Inquire For Price
Log in for availability
1 * 4 ST
Product Details & Documents

ETFE, white. Durable forceps for substrate handling.

  • Different tip styles available
  • Packaged in protective cases
Specifications
  • Type:
    Straight, 0,3 mm thick tip
  • Length:
    124 mm
  • Seq:
    01