ContactUsLinkComponent
ClearKlens First F7764 1L Ready-to-use sterile disinfectant Formulated with biguanide and quaternary ammonium Diluted with European pharmacopeia water Filtered to 0,2µm, irradiated, double bagged 10 x 1 litre = 1 carton 1 * 10 Bottle
  DIVYF7764
 :  Diversey
ClearKlens First F7764 1L Ready-to-use sterile disinfectant Formulated with biguanide and quaternary ammonium Diluted with European pharmacopeia water Filtered to 0,2µm, irradiated, double bagged 10 x 1 litre = 1 carton 1 * 10 Bottle
  DIVYF7764
 :  Diversey
Frequently Bought Together