ContactUsLinkComponent
Kit Master Mix PCR Phire Plant Direct 100 x 50 µl rxns 1 * 1 KIT
Catalog # F-160S
Supplier:  FINNZYMES REAGENTS
undefined
Kit Master Mix PCR Phire Plant Direct 100 x 50 µl rxns 1 * 1 KIT
Catalog # F-160S
Supplier:  FINNZYMES REAGENTS
Frequently Bought Together