Ponceau S
Glass bottle for solids, 50 g
Catalog # CAYM14330-50
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
- CAS Number:6226-79-5
- Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
- MDL Number:MFCD00003892
- Molecular Weight:760.56 g/mol
- Storage Temperature:Ambient
- EINECS:228-319-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



