Low Ionic Strength pH BufferSOLUTION pH6.96 ±0.04 @20°C 1 * 5 L
Catalog # REGELS695
Supplier: REAGECON
Low Ionic Strength pH BufferSOLUTION pH6.96 ±0.04 @20°C 1 * 5 L
Catalog # REGELS695
Supplier: REAGECON
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
UV/Visible standards, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Sulphuric acid 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1086804
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
VWR®, Turbidity standards, formazin
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
pH buffer solutions, colour coded
Supplier: Hach