SMZ745T Trinocular Stereo Zooming Body Optical System: Greenough type (zooming type) Trinocular Zoom ratio: 7,5:1 Zoom range: 3,35-300x (depending on eyepieces and auxiliary objective used) Tube: Fixed tube, Eyepiece inclination 45 Interpu 1 * 1 items
Catalog # NIKOMMA36410
Supplier: NIKON
Frequently Bought Together