Orders LinkContactUsLinkComponent
Digestion system, behrotest®
Digestion system, behrotest®
Complete crude fibre digestion unit with 4 sample positions, EXR 4
Artikelnr. BEHRB00218446
Anbieter:   BEHR
Digestion system, behrotest®
Artikelnr. BEHRB00218446
Anbieter:  BEHR
Digestion system, behrotest®
Digestion system, behrotest®
Complete crude fibre digestion unit with 4 sample positions, EXR 4
Preis
1 956,36 €
Log in for availability
1 * 1 ST
Product Details & Documents
Digestion apparatus for crude fibre or hydrolysis.
Delivery information: Includes serial heating unit with individually adjustable heating places, cooling water distribution with condenser rack and hoses, beakers (600 ml), cold fingers.
Specifications
  • Description:
    Complete crude fibre digestion unit with 4 sample positions
  • Seq:
    02
  • Type:
    EXR 4