Orders LinkContactUsLinkComponent
Digestion system, behrotest®
Digestion system, behrotest®
Complete crude fibre digestion unit with 6 sample positions, EXR 6
Catalog # BEHRB00218448
Supplier:   BEHR
Digestion system, behrotest®
Catalog # BEHRB00218448
Supplier:  BEHR
Digestion system, behrotest®
Digestion system, behrotest®
Complete crude fibre digestion unit with 6 sample positions, EXR 6
Price
2 435,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Digestion apparatus for crude fibre or hydrolysis.
 : Includes serial heating unit with individually adjustable heating places, cooling water distribution with condenser rack and hoses, beakers (600 ml), cold fingers.
Specifications
  • Description:
    Complete crude fibre digestion unit with 6 sample positions
  • Seq:
    03
  • Type:
    EXR 6