Flasks, round bottom, with three parallel necks
25 ml, 14/23 NS, 14/23 NS
Catalog # NEUB0760-0025-14
Supplier: NEUBERT VOLUME GLASSWAERE
Flasks, round bottom, with three parallel necks
Catalog # NEUB0760-0025-14
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Specifications
- Capacity:25 ml
- Seq:01
- Side necks:14/23 NS
- Centre neck:14/23 NS