Order Entry
Austria
Orders LinkContactUsLinkComponent
Brushes, FlexBrush™

Brushes, FlexBrush™

J-Brush adapter, hook-shaped brush, stainless steel with plastic handle, Nylon, 57,1 mm, Plastic, 406 mm
Catalog # 129-4202
   Waste Reducing
Supplier:   MICRONOVA
Brushes, FlexBrush™
Catalog # 129-4202
Supplier:  MICRONOVA
   Waste Reducing
Brushes, FlexBrush™
Brushes, FlexBrush™

J-Brush adapter, hook-shaped brush, stainless steel with plastic handle, Nylon, 57,1 mm, Plastic, 406 mm

Price
77,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

The FlexBrush™ is ideal for cleaning over and around piping and fittings. The flexibility of the head allows the user to shape the head for custom cleaning applications.

  • Bendable brush with polyester or microfibre cover
  • Acme threaded end attaches to a variety of handles for extended reach
  • Threaded handle (STA series) can be added for extended reach
  • Covers are compatible with strong disinfectants and autoclavable
Specifications
  • Bristle material:
    Nylon
  • Brush head Ø:
    57,1 mm
  • Handle material:
    Plastic
  • Description:
    J-Brush adapter, hook-shaped brush, stainless steel with plastic handle
  • Total length:
    406 mm
  • Cat. No.:
    129-4202
  • Environmentally Preferable:
    Waste Reducing
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.