Orders LinkContactUsLinkComponent
Test charts
Test charts
Test charts, White/black, 5C
Catalog # SIMEVF3006
Supplier:   Industrial Physics
Test charts
Catalog # SIMEVF3006
Supplier:  Industrial Physics
Test charts
Test charts
Test charts, White/black, 5C
Price
231,00 €
Log in for availability
1 * 250 ST
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
  • Dimensions from A6 up to A4
  • Film laminated for an excellent solvent, chemical resistance, and film spread
  • Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Ordering information: A certificate is available for charts with an optical brightener.
Specifications
  • Format:
    5C
  • Colour:
    White/black
  • Description:
    Test charts
  • Seq:
    15