Density hydrometers
Completely set with 25 instruments (00 to 22, incl. range finder), 160 mm
Catalog # AMARH800060
Supplier: Amarell
Springe zu:
Product Details & Documents
General purpose hydrometers for density areas.
- Paper scale and shot weighted body
- Hydrometer accuracy ±1 scale division
- Thermometer inside the hydrometer body
Used in laboratories and industry.
Spezifikationen
- Description:Completely set with 25 instruments (00 to 22, incl. range finder)
- Length:160 mm
- Seq:87