ContactUsLinkComponent
HayeSep A, 80/100, preconditioned, 41.28 mm coil, 5.58 feet (1.7 m) x 1/16 inch, 1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Catalog # HEWLG3591-81213
Supplier:  Agilent
HayeSep A, 80/100, preconditioned, 41.28 mm coil, 5.58 feet (1.7 m) x 1/16 inch, 1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Catalog # HEWLG3591-81213
Supplier:  Agilent
Frequently Bought Together