ContactUsLinkComponent
HayeSep A, 80/100, preconditioned, 41.28 mm coil, 2 feet (0.61 m) x 1/16 inch, 1 mm, UltiMetal, packed GC column, for Large Valve Ov en 1 * 1 items
Catalog # HEWLG3591-81212
Supplier:  Agilent
undefined
HayeSep A, 80/100, preconditioned, 41.28 mm coil, 2 feet (0.61 m) x 1/16 inch, 1 mm, UltiMetal, packed GC column, for Large Valve Ov en 1 * 1 items
Catalog # HEWLG3591-81212
Supplier:  Agilent
Frequently Bought Together