Wipro antibacterial red 1 * 20 items
Catalog # VILE137003
Supplier: FHP VILEDA SCS
Frequently Bought Together
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Cleanroom wipes, TechniCloth®
Supplier: Texwipe
VWR® Autoclavable Bags, Polypropylene, Clear
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
(1R,2R,3R,5S)-(-)-Isopinocampheylamine 95%
Supplier: Thermo Fisher Scientific
Disposable cannulas
Supplier: EHRHARDT SOEHNE
VWR® Spark-Free Freezers, –25 ℃, ATEX Certified
Supplier: VWR Collection
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies