BUFFER SOLUTION PH PH=4,01 25°C 1 * 1 l
Catalog # METN51340228
Supplier: Mettler - Toledo
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR® AVS TITRINORM® Potassium Chloride Electrolytic Solutions
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
Media additives and supplements
Supplier: Merck
Disposal Bags
Supplier: Brand
Buffer solutions in sachets
Supplier: Mettler - Toledo
Qualitix® Sterilisation Pouches
Supplier: Socorex Isba
Containers, wide mouth, square with thread
Supplier: Kautex
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Aquaresist Water Bath Protective Agent
Supplier: VWR Chemicals
SPRÜHKOPF F.500/1000ML BODE GEBINDE 1St 1 * 1 ST
Supplier: Roth Carl
ESD occupational shoes, clogs, C-FIT, Mia
Supplier: ELTEN
Dispensers for aluminium discs
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Accessories for Anaerobic Systems, Oxoid
Supplier: THERMO FISHER DIAGNOSTICS
pH buffer solutions
Supplier: Mettler - Toledo