ContactUsLinkComponent
Multipoint calibration set for Sievers 5310C, consisting of 1 x calibration blank (RTOCW), 1 each of 0.25mg/L (RTOCK15), 1mg/L (RTOC K09), 5mg/L (RTOCK14), 25mg/L (RTOCK12), 50mg/L (RTOCK13) C as KHP TOC standards and 1 each of 1mg/L (RTICN02), 5mg/L 1 * 40 mL
  REGERC120016
 :  REAGECON
Multipoint calibration set for Sievers 5310C, consisting of 1 x calibration blank (RTOCW), 1 each of 0.25mg/L (RTOCK15), 1mg/L (RTOC K09), 5mg/L (RTOCK14), 25mg/L (RTOCK12), 50mg/L (RTOCK13) C as KHP TOC standards and 1 each of 1mg/L (RTICN02), 5mg/L 1 * 40 mL
  REGERC120016
 :  REAGECON
Frequently Bought Together