ContactUsLinkComponent
5mg C/L Carbon Standard Set consisting of 1 x 40ml vial each of calibration blank (RTOCW), 5mg/L (RTOCK14) C TOC as KHP and 5 mg/L ( RTCIN05) C TIC as Na2CO3 1 * 40 mL
Catalog # REGERC120005
Supplier:  REAGECON
5mg C/L Carbon Standard Set consisting of 1 x 40ml vial each of calibration blank (RTOCW), 5mg/L (RTOCK14) C TOC as KHP and 5 mg/L ( RTCIN05) C TIC as Na2CO3 1 * 40 mL
Catalog # REGERC120005
Supplier:  REAGECON
Frequently Bought Together