ContactUsLinkComponent
1mg C/L Carbon Standard Set consisting of 1 x 40ml vial each of calibration blank (ROTCW), 1mg/L (RTOCK09) C TOC as KHP and 1 mg/L (RTICN02) C TIC as Na2CO3 1 * 40 mL
Catalog # REGERC120002
Supplier:  REAGECON
1mg C/L Carbon Standard Set consisting of 1 x 40ml vial each of calibration blank (ROTCW), 1mg/L (RTOCK09) C TOC as KHP and 1 mg/L (RTICN02) C TIC as Na2CO3 1 * 40 mL
Catalog # REGERC120002
Supplier:  REAGECON
Frequently Bought Together