Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Osmolality standards, protein based

Protein based osmolality standard, 280 mOsm/kg H₂O, Ampoule, 12× 5 ml, 1 KIT
Catalog # REGERE-POSM-280
Supplier:   REAGECON
Osmolality standards, protein based
Catalog # REGERE-POSM-280
Supplier:  REAGECON
CAS Number:  
Osmolality standards, protein based

Protein based osmolality standard, 280 mOsm/kg H₂O, Ampoule, 12× 5 ml, 1 KIT

CAS Number:  
Price
322,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Osmolality standards provide ease of use when calibrating all types of osmometers.

  • Ready to use and presented in convenient ampoules
  • Extended shelf life and has low uncertainty of measurement
  • Consistency of product-Independent, traceable
  • Consist of 12 ampoules of 5 ml

The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.

 : ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.

Specifications

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1 KIT
  • Description:
    Protein based osmolality standard
  • Pk Info:
    12× 5 ml
  • Osmolality:
    280 mOsm/kg H₂O

Specifications