Orders LinkContactUsLinkComponent
Inserts
Inserts
E 430/1 insert, 1/3 mesh tray, for various instruments, wire gauge with mesh size 5 mm, for upper and lower basket, G 7881, G 7891, 150×445×40 mm
Catalog # 141-0525
Supplier:   MIELE
Inserts
Catalog # 141-0525
Supplier:  MIELE
Inserts
Inserts
E 430/1 insert, 1/3 mesh tray, for various instruments, wire gauge with mesh size 5 mm, for upper and lower basket, G 7881, G 7891, 150×445×40 mm
Price
177,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    E 430/1 insert, 1/3 mesh tray, for various instruments, wire gauge with mesh size 5 mm, for upper and lower basket
  • Seq:
    14
  • W×D×H:
    150×445×40 mm
  • For:
    G 7881, G 7891