ContactUsLinkComponent
Desiccator wall antistatic, conductive polycarbonat/aluminium, with connecting thread (m6) for grounding, 6 separate chambers, each chamber with: 1 elastic gasket-lined door with magnetic lock, 1 desiccant tray, 1 mounted hygrometer, 1 removable shel 1 * 1 items
Catalog # BOHLV1997-08
Supplier:  Bohlender
undefined
Desiccator wall antistatic, conductive polycarbonat/aluminium, with connecting thread (m6) for grounding, 6 separate chambers, each chamber with: 1 elastic gasket-lined door with magnetic lock, 1 desiccant tray, 1 mounted hygrometer, 1 removable shel 1 * 1 items
Catalog # BOHLV1997-08
Supplier:  Bohlender
Frequently Bought Together