ContactUsLinkComponent
Big-star-desiccator antistatic, conductive polycarbonate/aluminium, 1 chamber, with: 1 elastic gasket-lined door, each 1 desiccant t ray, each 1 mounted hygrometer, 2 removable shelves made of stainless steel, 1 connecting thread (m6) for grounding. 1 * 1 items
Catalog # BOHLV1925-07
Supplier:  Bohlender
undefined
Big-star-desiccator antistatic, conductive polycarbonate/aluminium, 1 chamber, with: 1 elastic gasket-lined door, each 1 desiccant t ray, each 1 mounted hygrometer, 2 removable shelves made of stainless steel, 1 connecting thread (m6) for grounding. 1 * 1 items
Catalog # BOHLV1925-07
Supplier:  Bohlender
Frequently Bought Together