Order Entry
Austria
Orders LinkContactUsLinkComponent
pH buffer solutions, colour coded

pH buffer solutions, colour coded

Buffer solution, 7,00, ±0,02, Phosphate, Yellow, 25 °C, 50 mL
Catalog # HACH2283526
Supplier:   Hach
pH buffer solutions, colour coded
Catalog # HACH2283526
Supplier:  Hach
CAS Number:  
pH buffer solutions, colour coded
pH buffer solutions, colour coded

Buffer solution, 7,00, ±0,02, Phosphate, Yellow, 25 °C, 50 mL

CAS Number:  
Price
14,10 €
Log in for availability
1 * 50 mL
Product Details & Documents

For calibration of pH meters

  • Colour coded for easy identification
  • Accuracy (pH): ±0,02 at 25 °C

 : Traceable to NIST.

Specifications

Specifications

  • pH buffer type:
    Phosphate
  • Pk:
    50 mL
  • Reference temperature:
    25 °C
  • Colour:
    Yellow
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,02
  • pH value:
    7,00

Specifications