ContactUsLinkComponent
RLC meter, wide test frequency 20Hz 10MHz, 0.1% basic accuracy and 6 digits measurement, large lCD display with intuitive user int erface, full measuring functions with dut V/I monitor. 1 * 1 items
Catalog # DIDAPMD400130
Supplier:  DIDALAB
RLC meter, wide test frequency 20Hz 10MHz, 0.1% basic accuracy and 6 digits measurement, large lCD display with intuitive user int erface, full measuring functions with dut V/I monitor. 1 * 1 items
Catalog # DIDAPMD400130
Supplier:  DIDALAB
Frequently Bought Together