ContactUsLinkComponent
Multimeter, DC voltage: 100mv~1000v, AC voltage: 100mv~750v, DC current : 100µa-10a, AC current: 1ma-10a, R 100 ohms~100 mohms, RTD -200°c~600°c, thermocouple -200°c~1820°c, 6 ½ digit display : 1,200,000 counts, DCV basic accuracy 1 * 1 items
Catalog # DIDAPMD400100
Supplier:  DIDALAB
Multimeter, DC voltage: 100mv~1000v, AC voltage: 100mv~750v, DC current : 100µa-10a, AC current: 1ma-10a, R 100 ohms~100 mohms, RTD -200°c~600°c, thermocouple -200°c~1820°c, 6 ½ digit display : 1,200,000 counts, DCV basic accuracy 1 * 1 items
Catalog # DIDAPMD400100
Supplier:  DIDALAB
Frequently Bought Together