ContactUsLinkComponent
Power supply, multiple, programmable, voltage CH1 0/30V, CH2 0/30V, CH3 2.5/3.3/5.0V, current 0-3A, 3 independent isolated outputs, 4 led display sets 3 digits after decimal point, minimum resolution 1MV/1MA, digital panel co 1 * 1 items
Catalog # DIDAPMD300020_UK
Supplier:  DIDALAB
Power supply, multiple, programmable, voltage CH1 0/30V, CH2 0/30V, CH3 2.5/3.3/5.0V, current 0-3A, 3 independent isolated outputs, 4 led display sets 3 digits after decimal point, minimum resolution 1MV/1MA, digital panel co 1 * 1 items
Catalog # DIDAPMD300020_UK
Supplier:  DIDALAB
Frequently Bought Together