ContactUsLinkComponent
Oscilloscope, digital, bandwidth 150MHz, 2 channels, visual persistence oscilloscope, 8 inches screen, 800x600 high resolution TFT L CD didplay, built-in 3 input impedance selection, 50ohms/75ohms/1mega ohms. 1 * 1 items
Catalog # 761-1000
Supplier:  DIDALAB
undefined
Oscilloscope, digital, bandwidth 150MHz, 2 channels, visual persistence oscilloscope, 8 inches screen, 800x600 high resolution TFT L CD didplay, built-in 3 input impedance selection, 50ohms/75ohms/1mega ohms. 1 * 1 items
Catalog # 761-1000
Supplier:  DIDALAB
Frequently Bought Together