ContactUsLinkComponent
Filter interference, 578nm, yellow, diameter 40mm, made of thin layer deposits, extremely selective filter around an accurate wavele ngth, supplied with componenet holder and individual calibration, tolerance +/-2nm, width 10nm, min 1 * 1 items
Catalog # DIDAPOD010573
Supplier:  DIDALAB
Filter interference, 578nm, yellow, diameter 40mm, made of thin layer deposits, extremely selective filter around an accurate wavele ngth, supplied with componenet holder and individual calibration, tolerance +/-2nm, width 10nm, min 1 * 1 items
Catalog # DIDAPOD010573
Supplier:  DIDALAB
Frequently Bought Together