ContactUsLinkComponent
Filter interference, 633nm, red, diameter 40mm, made of thin layer deposits, extremely selective filter around an accurate wavelengt h, supplied with componenet holder and individual calibration, tolerance +/-2 nm, width 10nm, min 1 * 1 items
Catalog # DIDAPOD010574
Supplier:  DIDALAB
Filter interference, 633nm, red, diameter 40mm, made of thin layer deposits, extremely selective filter around an accurate wavelengt h, supplied with componenet holder and individual calibration, tolerance +/-2 nm, width 10nm, min 1 * 1 items
Catalog # DIDAPOD010574
Supplier:  DIDALAB
Frequently Bought Together